| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700010657.mm.1 |
TC0700001203.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700010657.mm.1 |
NM_023403.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010657.mm.1 |
NM_023403.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010656.mm.1 |
TC0700001203.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700010656.mm.1 |
NM_023403.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010656.mm.1 |
NM_023403.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010659.mm.1 |
TC0700001203.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700010659.mm.1 |
NM_023403.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010659.mm.1 |
NM_023403.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010658.mm.1 |
TC0700001203.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700010658.mm.1 |
NM_023403.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700010658.mm.1 |
NM_023403.3 |
Refseq |