Assay Details
| Gene Symbol: Tfpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290381.1 | NP_001277310.1 | 3 - 4 | 711 | 80 | |
| NM_023524.2 | NP_076013.1 | 3 - 4 | 711 | 80 | ||
| GenBank mRNA | AB035134.1 | - | 3 - 4 | 402 | 80 | |
| AF267988.1 | - | 3 - 4 | 666 | 80 | ||
| AK010279.1 | - | 3 - 4 | 369 | 80 | ||
| AK155925.1 | - | 3 - 4 | 695 | 80 | ||
| BC024356.1 | - | 3 - 4 | 610 | 5066740 | 80 | |
| CX768547.1 | - | 2 - 3 | 230 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tfpt
Genomic Map