Assay Details
Gene Symbol: Fbxo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081490.2 | NP_001074959.1 | 10 - 11 | 1017 | 62 | |
NM_023605.2 | NP_076094.2 | 10 - 11 | 1319 | 62 | ||
GenBank mRNA | AB041585.1 | - | 10 - 11 | 1289 | 62 | |
AK077607.1 | - | 10 - 11 | 980 | 62 | ||
BC020074.1 | - | 10 - 11 | 1290 | 3710611 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
