Assay Details
| Gene Symbol: Rab27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301230.1 | NP_001288159.1 | 6 - 7 | 841 | 61 | |
| NM_001301232.1 | NP_001288161.1 | 5 - 6 | 734 | 61 | ||
| NM_023635.6 | NP_076124.1 | 5 - 6 | 692 | 61 | ||
| GenBank mRNA | AB046693.1 | - | 4 - 5 | 490 | 61 | |
| AB120511.1 | - | 4 - 5 | 468 | 61 | ||
| AB232622.1 | - | 4 - 5 | 468 | 61 | ||
| AF304376.1 | - | 5 - 6 | 647 | 61 | ||
| AK010373.1 | - | 5 - 6 | 610 | 61 | ||
| AK030529.1 | - | 5 - 6 | 639 | 61 | ||
| BC008173.1 | - | 6 - 7 | 798 | 3710213 | 61 | |
| CV784419.1 | - | 5 - 6 | 603 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab27a
Genomic Map