Assay Details
| Gene Symbol: Kdelc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023645.3 | NP_076134.1 | 5 - 6 | 1118 | 63 | |
| GenBank mRNA | AJ404004.1 | - | 5 - 6 | 880 | 63 | |
| AK007835.1 | - | 5 - 6 | 1111 | 63 | ||
| AK035899.1 | - | 5 - 6 | 1119 | 63 | ||
| AK053315.1 | - | 5 - 6 | 1111 | 63 | ||
| AK079701.1 | - | 5 - 6 | 1111 | 63 | ||
| AK170300.1 | - | 5 - 6 | 1147 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kdelc1
Genomic Map