Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800004516.mm.1 |
NM_001252620.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004516.mm.1 |
NM_001252620.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004516.mm.1 |
NM_001252621.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004516.mm.1 |
NM_001252621.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004516.mm.1 |
NM_023689.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004516.mm.1 |
NM_023689.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004516.mm.1 |
TC0800000661.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800004517.mm.1 |
NM_001252620.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004517.mm.1 |
NM_001252620.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004517.mm.1 |
NM_001252621.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004517.mm.1 |
NM_001252621.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004517.mm.1 |
NM_023689.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004517.mm.1 |
NM_023689.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800004517.mm.1 |
TC0800000661.mm.1 |
MTA TC |