Assay Details
| Gene Symbol: Abcb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023732.2 | NP_076221.1 | 1 - 2 | 717 | 74 | |
| GenBank mRNA | AK004605.1 | - | 1 - 2 | 797 | 74 | |
| AK040323.1 | - | 1 - 2 | 721 | 74 | ||
| AK150853.1 | - | 1 - 2 | 805 | 74 | ||
| AK151165.1 | - | 1 - 2 | 826 | 74 | ||
| AK152584.1 | - | 1 - 2 | 805 | 74 | ||
| AK164003.1 | - | 1 - 2 | 721 | 74 | ||
| AK168642.1 | - | 1 - 2 | 818 | 74 | ||
| BC006634.1 | - | 1 - 2 | 717 | 3710552 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcb6
Genomic Map