Assay Details
| Gene Symbol: Abcb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023732.2 | NP_076221.1 | 14 - 15 | 2135 | 118 | |
| GenBank mRNA | AK004605.1 | - | 14 - 15 | 2215 | 118 | |
| AK151165.1 | - | 14 - 15 | 2244 | 118 | ||
| AK168642.1 | - | 14 - 15 | 2236 | 118 | ||
| AK200467.1 | - | 3 - 4 | 206 | 118 | ||
| AK208919.1 | - | 3 - 4 | 206 | 118 | ||
| AK210369.1 | - | 1 - 2 | 80 | 118 | ||
| AK212610.1 | - | 3 - 4 | 206 | 118 | ||
| BC006634.1 | - | 14 - 15 | 2135 | 3710552 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcb6
Genomic Map