Assay Details
Gene Symbol: Fkbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024169.3 | NP_077131.2 | 2 - 3 | 306 | 77 | |
GenBank mRNA | AK003331.1 | - | 2 - 3 | 307 | 77 | |
AK007813.1 | - | 2 - 3 | 270 | 77 | ||
BC002311.1 | - | 2 - 3 | 260 | 77 | ||
BC022900.1 | - | 2 - 3 | 260 | 77 | ||
BC037596.1 | - | 2 - 3 | 262 | 5250344 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
