Assay Details
| Gene Symbol: Fkbp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024169.3 | NP_077131.2 | 3 - 4 | 397 | 67 | |
| GenBank mRNA | AK003331.1 | - | 3 - 4 | 398 | 67 | |
| AK007813.1 | - | 3 - 4 | 361 | 67 | ||
| AK019132.1 | - | 3 - 4 | 339 | 67 | ||
| BC002311.1 | - | 3 - 4 | 351 | 67 | ||
| BC037596.1 | - | 3 - 4 | 353 | 5250344 | 67 | |
More Information
Set Membership: |
|

Fkbp11
Genomic Map