Assay Details
Gene Symbol: Cmtm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024217.3 | NP_077179.1 | 4 - 5 | 676 | 76 | |
GenBank mRNA | AK003882.1 | - | 4 - 5 | 532 | 76 | |
AK079165.1 | - | 4 - 5 | 645 | 76 | ||
AK172091.1 | - | 4 - 5 | 579 | 76 | ||
AY241870.1 | - | 4 - 5 | 670 | 76 | ||
BC003230.1 | - | 4 - 5 | 583 | 3483351 | 76 | |
BC145817.1 | - | 4 - 5 | 548 | 40131120 | 76 | |
BC145819.1 | - | 4 - 5 | 548 | 40131122 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26127987 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26127991 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26127989 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26127992 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26127993 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321886 | AK079165.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321886 | AK172091.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321886 | AY241870.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321886 | BC003230.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321886 | NM_024217.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321886 | AK079165.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321886 | AK172091.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321886 | AY241870.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321886 | BC003230.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321886 | NM_024217.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512752 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176722 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892284 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178707 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321886 | AK079165.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321886 | AK172091.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321886 | AY241870.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321886 | BC003230.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321886 | NM_024217.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321886 | AK079165.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321886 | AK172091.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321886 | AY241870.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321886 | BC003230.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321886 | NM_024217.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321886 | AK079165.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321886 | AK172091.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321886 | AY241870.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321886 | BC003230.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321886 | NM_024217.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321886 | AK079165.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321886 | AK172091.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321886 | AY241870.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321886 | BC003230.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321886 | NM_024217.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127969 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26128071 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127991 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26128073 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127966 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26128006 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26128001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127986 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127990 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127989 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26128007 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127984 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127976 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127977 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127970 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127996 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127987 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127979 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127974 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26128005 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26128074 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127972 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26128070 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127982 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127983 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127992 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127985 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26128002 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26128072 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127975 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127998 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178707 | - | - |
Whole Mouse Genome G4122A | A_51_P321886 | AK079165.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P321886 | AK172091.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P321886 | AY241870.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P321886 | BC003230.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P321886 | NM_024217.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P321886 | AK079165.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P321886 | AK172091.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P321886 | AY241870.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P321886 | BC003230.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P321886 | NM_024217.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102987 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178707 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07022424 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07022408 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07022416 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07448873 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07448875 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26127993 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26127989 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26127992 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26127987 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26127991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26128064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07004191 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128012 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127998 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128060 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128026 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128014 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127968 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128009 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128039 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127975 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127971 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128036 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127977 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127978 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127976 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128051 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07011551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06860502 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127996 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127963 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07022427 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127989 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127959 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06860512 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06840180 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07231999 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127981 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128030 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127960 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07232016 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07232011 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128007 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128008 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127972 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128038 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07022415 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07011556 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128055 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127995 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07011554 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128073 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128045 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128033 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07022413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127957 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127961 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127966 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07441573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07022421 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07232018 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128022 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26128023 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127956 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26127987 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07232003 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10574476 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574473 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574474 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574472 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574475 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574477 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10574471 | - | - |
Mouse Genome 430 2.0 Array | 1434337_at | - | - |
Mouse Genome 430 2.0 Array | 1448316_at | - | - |
Mouse Genome 430 2.0 Array | 1448316_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1434337_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68119_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68119_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800009190.mm.1 | NM_024217.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800009190.mm.1 | NM_024217.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800009190.mm.1 | TC0800001182.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800009191.mm.1 | NM_024217.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800009191.mm.1 | NM_024217.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800009191.mm.1 | TC0800001182.mm.1 | MTA TC |
Mouse Expression Set 430 | 1448316_at | - | - |
Mouse Expression Set 430 | 1448316_PM_at | - | - |
Murine Genome U74v2 Set | 97360_at | - | - |
Murine Genome U74v2 Set | 97360_PM_at | - | - |
Murine Genome U74v2 Set | 169552_PM_r_at | - | - |
Murine Genome U74v2 Set | 169552_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1448316_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448316_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 919247 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1940180 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1940180 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001940180 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2679037 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2679037 | - | - |
mouse-6 v1.1 (Illumina) | scl33440.8_211-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1940180 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl33440.8_211-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1940180 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2679037 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2679037 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl33440.8_211-S | - | - |
