Assay Details
Gene Symbol: Pcyt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024229.2 | NP_077191.2 | 9 - 10 | 849 | 77 | |
GenBank mRNA | AK140458.1 | - | 9 - 10 | 889 | 77 | |
AK144544.1 | - | 9 - 10 | 898 | 77 | ||
AK166319.1 | - | 8 - 9 | 824 | 77 | ||
BC003473.1 | - | 9 - 10 | 855 | 2650077 | 77 | |
BC008276.1 | - | 9 - 10 | 849 | 3707732 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
