Assay Details
| Gene Symbol: Tmem79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024246.5 | NP_077208.1 | 2 - 3 | 846 | 89 | |
| GenBank mRNA | AK009760.1 | - | 2 - 3 | 847 | 89 | |
| AK010144.1 | - | 2 - 3 | 936 | 89 | ||
| BC003309.1 | - | 2 - 3 | 901 | 3154669 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem79
Genomic Map