Assay Details
Gene Symbol: Ergic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026170.3 | NP_080446.1 | 1 - 2 | 123 | 56 | |
GenBank mRNA | AK003239.1 | - | 1 - 2 | 110 | 56 | |
AK004629.1 | - | 1 - 2 | 77 | 56 | ||
AK084385.1 | - | 1 - 2 | 70 | 56 | ||
AK088157.1 | - | 1 - 2 | 78 | 56 | ||
AK151239.1 | - | 1 - 2 | 77 | 56 | ||
AK173123.1 | - | 1 - 2 | 112 | 56 | ||
BC005516.1 | - | 1 - 2 | 123 | 3494155 | 56 | |
BC083144.1 | - | 1 - 2 | 43 | 30432732 | 56 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK004629.1,AK084385.1,AK088157.1,AK151239.1,BC005516.1,BC083144.1,NM_026170.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
