Assay Details
Gene Symbol: Ergic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026170.3 | NP_080446.1 | 6 - 7 | 586 | 112 | |
GenBank mRNA | AB093571.1 | - | 6 - 7 | 485 | 112 | |
AK003239.1 | - | 6 - 7 | 572 | 112 | ||
AK004629.1 | - | 6 - 7 | 540 | 112 | ||
AK040727.1 | - | 3 - 4 | 738 | 112 | ||
AK084385.1 | - | 6 - 7 | 533 | 112 | ||
AK088157.1 | - | 6 - 7 | 541 | 112 | ||
AK151239.1 | - | 6 - 7 | 540 | 112 | ||
AK173123.1 | - | 6 - 7 | 575 | 112 | ||
AK194494.1 | - | 1 - 2 | 55 | 112 | ||
AK196223.1 | - | 1 - 2 | 55 | 112 | ||
AK196818.1 | - | 1 - 2 | 55 | 112 | ||
AK202556.1 | - | 1 - 2 | 55 | 112 | ||
AK204170.1 | - | 1 - 2 | 55 | 112 | ||
BC005516.1 | - | 6 - 7 | 586 | 3494155 | 112 | |
BC083144.1 | - | 6 - 7 | 506 | 30432732 | 112 | |
CT010230.1 | - | 6 - 7 | 485 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
