Assay Details
Gene Symbol: Glipr1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026223.2 | NP_080499.1 | 1 - 2 | 243 | 75 | |
GenBank mRNA | AK014846.1 | - | 1 - 2 | 244 | 75 | |
AK015834.1 | - | 1 - 2 | 227 | 75 | ||
AK161292.1 | - | 1 - 2 | 244 | 75 | ||
BC118017.1 | - | 1 - 2 | 244 | 40102576 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P263130 | AK014846.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P263130 | AK015834.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P263130 | AK161292.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P263130 | NM_026223.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P263130 | AK014846.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P263130 | AK015834.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P263130 | AK161292.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P263130 | NM_026223.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916950 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281682 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972214 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704224 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84960 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578604 | AK018428.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578604 | AK039348.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578604 | AK039401.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578604 | AK155128.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578604 | AK220395.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578604 | BC003918.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578604 | BC025010.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578604 | BC038002.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578604 | BC052213.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578604 | BC079676.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578604 | NM_028982.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323929 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396863 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 542148 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 129697 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151838 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163642 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143587 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129295 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P263130 | AK014846.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P263130 | AK015834.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P263130 | AK161292.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P263130 | NM_026223.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P263130 | AK014846.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P263130 | AK015834.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P263130 | AK161292.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P263130 | NM_026223.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452467 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452465 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452460 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452481 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452480 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452475 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452455 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452452 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452477 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452458 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452471 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452468 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452463 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452474 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452470 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27452461 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07194801 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129295 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143587 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07194781 | - | - |
Whole Mouse Genome G4122A | A_52_P263130 | AK014846.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P263130 | AK015834.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P263130 | AK161292.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P263130 | NM_026223.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P263130 | AK014846.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P263130 | AK015834.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P263130 | AK161292.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P263130 | NM_026223.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113955 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143587 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129295 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08320591 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08320599 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08839935 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08839941 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08320595 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08839939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27452468 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452454 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452461 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452455 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452477 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08320599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452456 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452433 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452460 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08290281 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452431 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08565887 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452428 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452434 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452463 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452506 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452480 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08320594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08839938 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452470 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452452 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452473 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08599840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27452475 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10366369 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10366373 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10366374 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10366371 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10366370 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10366372 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10366375 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419733_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10366368 | - | - |
Mouse Genome 430 2.0 Array | 1419733_at | - | - |
Mouse Genome 430 2.0 Array | 1419733_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67537_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67537_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1000009766.mm.1 | NM_026223.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000009766.mm.1 | NM_026223.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000009766.mm.1 | TC1000001306.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000009767.mm.1 | NM_026223.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000009767.mm.1 | NM_026223.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000009767.mm.1 | TC1000001306.mm.1 | MTA TC |
Mouse Expression Set 430 | 1419733_PM_at | - | - |
Mouse Expression Set 430 | 1419733_at | - | - |
Murine Genome U74v2 Set | 166231_at | - | - |
Murine Genome U74v2 Set | 166231_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419733_at | - | - |
Mouse Genome 430A 2.0 Array | 1419733_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419733_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419733_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 308179 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1410044 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1410044 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001410044 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2654176 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2654176 | - | - |
mouse-6 v1.1 (Illumina) | scl38440.6.1_288-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1410044 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl38440.6.1_288-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1410044 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2654176 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2654176 | - | - |
