Assay Details
Gene Symbol: Larp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026235.4 | NP_080511.2 | 1 - 2 | 291 | 108 | |
GenBank mRNA | AK017372.1 | - | 1 - 2 | 296 | 108 | |
AK020966.1 | - | 1 - 2 | 228 | 108 | ||
AK039614.1 | - | 1 - 2 | 274 | 108 | ||
AK042616.1 | - | 1 - 2 | 223 | 108 | ||
AK087521.1 | - | 1 - 2 | 294 | 108 | ||
BC090615.1 | - | 1 - 2 | 423 | 6809572 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26560490 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26560493 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26560489 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26560491 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P390285 | AK017372.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P390285 | AK017372.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455494 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 942705 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962301 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 256834 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259306 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869611 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339498 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292661 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789883 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 448989 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 257332 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422541 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767295 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132651 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61813 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112957 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167278 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P390285 | AK017372.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P390285 | AK017372.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560469 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560490 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560473 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560489 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560496 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560506 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560502 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560486 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560493 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560478 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560504 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560508 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560483 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560479 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560499 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560474 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560509 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560476 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560503 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560484 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560487 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560491 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560497 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560500 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560498 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560471 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26560481 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167278 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112957 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01947576 | - | - |
Whole Mouse Genome G4122A | A_51_P390285 | AK017372.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P390285 | AK017372.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119613 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167278 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112957 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07943327 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07943324 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07722665 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07943321 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07722664 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07722666 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26560493 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26560491 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26560489 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26560490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26560618 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07689781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560521 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560497 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560505 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560496 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560486 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560506 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560478 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560479 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560487 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560510 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07689773 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07943324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07722666 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560508 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07701405 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560498 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560609 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07709351 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560613 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560509 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07502360 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560614 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560473 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560622 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560519 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560474 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560500 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560467 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560540 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560493 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07632436 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560526 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560491 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560515 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560618 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560522 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07722664 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07699020 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560472 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560464 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560623 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08018135 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560539 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560494 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07578478 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560476 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560537 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560538 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08158509 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560528 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560535 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26560483 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07578484 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10586014 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10586013 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10586012 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418400_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1443028_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10586011 | - | - |
Mouse Genome 430 2.0 Array | 1443028_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1443028_at | - | - |
Mouse Genome 430 2.0 Array | 1418400_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418400_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67557_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67557_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900005727.mm.1 | NM_026235.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900005727.mm.1 | NM_026235.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900005727.mm.1 | TC0900000767.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900005728.mm.1 | NM_026235.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900005728.mm.1 | NM_026235.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900005728.mm.1 | TC0900000767.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900005726.mm.1 | TC0900000767.mm.1 | MTA TC |
Mouse Expression Set 430 | 1418400_at | - | - |
Mouse Expression Set 430 | 1418400_PM_at | - | - |
Mouse Expression Set 430 | 1443028_PM_at | - | - |
Mouse Expression Set 430 | 1443028_at | - | - |
Murine Genome U74v2 Set | 139238_PM_at | - | - |
Murine Genome U74v2 Set | 139238_at | - | - |
Mouse Genome 430A 2.0 Array | 1418400_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418400_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418400_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418400_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443028_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443028_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 476672 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 580364 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 580364 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1570537 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000580364 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001570537 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219564 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2594779 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1219564 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2594779 | - | - |
mouse-6 v1.1 (Illumina) | scl0003530.1_1417-S | - | - |
mouse-6 v1.1 (Illumina) | scl067557.3_10-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1570537 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 580364 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003530.1_1417-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl067557.3_10-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 580364 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1570537 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2594779 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1219564 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219564 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2594779 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003530.1_1417-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067557.3_10-S | - | - |
