Assay Details
Gene Symbol: Cpeb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290676.1 | NP_001277605.1 | 5 - 6 | 2870 | 93 | |
NM_001290678.1 | NP_001277607.1 | 4 - 5 | 2819 | 93 | ||
NM_026252.4 | NP_080528.2 | 6 - 7 | 2894 | 93 | ||
GenBank mRNA | AK015381.1 | - | 3 - 4 | 289 | 93 | |
AK015401.1 | - | 2 - 3 | 277 | 93 | ||
AK173229.1 | - | 4 - 5 | 2828 | 93 | ||
AY313775.1 | - | 6 - 7 | 1624 | 93 | ||
BC079599.1 | - | 5 - 6 | 678 | 93 | ||
BC115430.1 | - | 4 - 5 | 1543 | 40094123 | 93 | |
BC115431.1 | - | 4 - 5 | 1543 | 40094124 | 93 | |
BC145863.1 | - | 6 - 7 | 1618 | 40131166 | 93 | |
BC145865.1 | - | 6 - 7 | 1618 | 40131168 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
