Assay Details
Gene Symbol: Slc25a26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026255.5 | NP_080531.2 | 8 - 9 | 747 | 78 | |
GenBank mRNA | AK015299.1 | - | 8 - 9 | 729 | 78 | |
AK015954.1 | - | 8 - 9 | 766 | 78 | ||
AK017037.1 | - | 11 - 12 | 1121 | 78 | ||
BC019156.1 | - | 9 - 10 | 717 | 5066084 | 78 | |
BC019170.1 | - | 8 - 9 | 748 | 5003084 | 78 | |
BC037142.1 | - | 8 - 9 | 724 | 5026631 | 78 | |
BC049899.1 | - | 8 - 9 | 884 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
