Assay Details
Gene Symbol: Tctn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026260.2 | NP_080536.2 | 2 - 3 | 411 | 96 | |
GenBank mRNA | AK015861.1 | - | 2 - 3 | 399 | 96 | |
AK016982.1 | - | 2 - 3 | 397 | 96 | ||
AK030156.1 | - | 2 - 3 | 397 | 96 | ||
BC027335.1 | - | 2 - 3 | 411 | 3710046 | 96 | |
BC080805.1 | - | 2 - 3 | 386 | 5352578 | 96 | |
DQ278871.1 | - | 2 - 3 | 428 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
