Assay Details
Gene Symbol: Tctn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026260.2 | NP_080536.2 | 5 - 6 | 778 | 59 | |
GenBank mRNA | AK015861.1 | - | 5 - 6 | 766 | 59 | |
AK016982.1 | - | 5 - 6 | 764 | 59 | ||
AK030156.1 | - | 5 - 6 | 764 | 59 | ||
BC027335.1 | - | 5 - 6 | 778 | 3710046 | 59 | |
BC080805.1 | - | 5 - 6 | 753 | 5352578 | 59 | |
DQ278871.1 | - | 5 - 6 | 795 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
