Assay Details
Gene Symbol: Tctn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026260.2 | NP_080536.2 | 9 - 10 | 1135 | 60 | |
GenBank mRNA | AK015861.1 | - | 9 - 10 | 1123 | 60 | |
AK016982.1 | - | 9 - 10 | 1121 | 60 | ||
BC027335.1 | - | 9 - 10 | 1135 | 3710046 | 60 | |
BC080805.1 | - | 9 - 10 | 1110 | 5352578 | 60 | |
DQ278871.1 | - | 9 - 10 | 1152 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
