Assay Details
Gene Symbol: Rpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026308.2 | NP_080584.1 | 3 - 4 | 247 | 100 | |
GenBank mRNA | AJ504717.1 | - | 3 - 4 | 254 | 100 | |
AK002780.1 | - | 3 - 4 | 248 | 100 | ||
AK003455.1 | - | 3 - 4 | 245 | 100 | ||
AY251016.1 | - | 3 - 4 | 269 | 100 | ||
AY251017.1 | - | 2 - 3 | 168 | 100 | ||
BC028491.1 | - | 3 - 4 | 257 | 1497187 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31163327 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31163283 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31163325 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31163284 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31163326 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31163324 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143337 | BC028491.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143337 | NM_026308.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143337 | AJ504717.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143337 | AK002780.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143337 | AK003455.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143336 | BC028491.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143336 | NM_026308.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143336 | AK002780.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143336 | AK003455.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2143336 | AY251016.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437886 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 68902 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151625 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143337 | BC028491.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143337 | NM_026308.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143337 | AJ504717.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143337 | AK002780.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143337 | AK003455.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143336 | BC028491.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143336 | NM_026308.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143336 | AK002780.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143336 | AK003455.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2143336 | AY251016.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178646 | NM_026308.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178646 | AJ504717.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178646 | AK002780.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178646 | AK003455.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P178646 | AY251016.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163318 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163334 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163325 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163316 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163328 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163322 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163329 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163319 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163315 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163313 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163326 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163324 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163317 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163330 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163332 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163320 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151625 | - | - |
Whole Mouse Genome G4122A | A_51_P178646 | NM_026308.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P178646 | AJ504717.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P178646 | AK002780.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P178646 | AK003455.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P178646 | AY251016.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105568 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151625 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13083395 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13513660 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13083397 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13083391 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13513665 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13083388 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31163326 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31163327 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31163325 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31163283 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31163284 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31163324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163395 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163330 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163285 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163281 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163328 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163303 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163319 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163313 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163326 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13239039 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13330149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13330153 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13513660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163289 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163321 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163311 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163307 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13083391 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163304 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163279 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13513668 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13291442 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163290 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163318 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163315 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163322 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163273 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163324 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13301105 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163329 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31163308 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10450747 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450748 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450750 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450751 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450749 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449021_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10450746 | - | - |
Mouse Genome 430 2.0 Array | 1449021_at | - | - |
Mouse Genome 430 2.0 Array | 1449021_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67676_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67676_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700019299.mm.1 | NM_026308.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019299.mm.1 | NM_026308.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019299.mm.1 | TC1700002017.mm.1 | MTA TC |
Mouse Expression Set 430 | 1449021_PM_at | - | - |
Mouse Expression Set 430 | 1449021_at | - | - |
Murine Genome U74v2 Set | 96618_at | - | - |
Murine Genome U74v2 Set | 96618_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449021_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449021_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449021_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449021_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 911759 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5890025 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005890025 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2726308 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2726308 | - | - |
mouse-6 v1.1 (Illumina) | scl49959.3.1_23-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5890025 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl49959.3.1_23-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5890025 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2726308 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2726308 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl49959.3.1_23-S | - | - |
