Assay Details
Gene Symbol: Lsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026309.2 | NP_080585.1 | 1 - 2 | 43 | 67 | |
GenBank mRNA | AK003125.1 | - | 1 - 2 | 46 | 67 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK003125.1,NM_026309.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24639527 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24639530 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24639528 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349023 | BC054368.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349023 | BC062875.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349023 | AK003125.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349023 | AK007849.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349023 | NM_026309.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349023 | BC054368.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349023 | BC062875.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349023 | AK003125.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349023 | AK007849.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P349023 | NM_026309.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292056 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819375 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20612 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 765500 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155929 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349023 | BC054368.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349023 | BC062875.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349023 | AK003125.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349023 | AK007849.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349023 | NM_026309.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349023 | BC054368.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349023 | BC062875.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349023 | AK003125.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349023 | AK007849.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P349023 | NM_026309.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P349023 | BC054368.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P349023 | BC062875.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P349023 | AK003125.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P349023 | AK007849.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P349023 | NM_026309.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P349023 | BC054368.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P349023 | BC062875.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P349023 | AK003125.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P349023 | AK007849.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P349023 | NM_026309.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639539 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639553 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639528 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639547 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639550 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639537 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639544 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639563 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639555 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639543 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639565 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639552 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639557 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639527 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639548 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639541 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639559 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639531 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639535 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639551 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639562 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639532 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639540 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639567 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639545 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24639530 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155929 | - | - |
Whole Mouse Genome G4122A | A_51_P349023 | BC054368.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349023 | BC062875.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349023 | AK003125.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349023 | AK007849.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349023 | NM_026309.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349023 | BC054368.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349023 | BC062875.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349023 | AK003125.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349023 | AK007849.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P349023 | NM_026309.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114301 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138104 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155929 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05878403 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05115305 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05878402 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05878404 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24639530 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24639528 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24639527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24639604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05778436 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639583 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05294050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05778433 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639579 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05878403 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05878402 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639627 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639547 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639611 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639537 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639640 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639575 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05878404 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639581 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05136799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05426950 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05637102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639572 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639534 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05381340 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639625 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639582 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05708587 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05778432 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05778431 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05492426 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639539 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05492420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05778435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639535 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639612 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05778430 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639528 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639556 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05381342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639525 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639626 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05426951 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639555 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639562 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24639573 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10540121 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10540119 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10540120 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448536_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10540118 | - | - |
Mouse Genome 430 2.0 Array | 1448536_at | - | - |
Mouse Genome 430 2.0 Array | 1448536_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67678_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67678_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600008519.mm.1 | TC0600001088.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600008519.mm.1 | NM_026309.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600008519.mm.1 | NM_026309.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600008521.mm.1 | TC0600001088.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600008521.mm.1 | NM_026309.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600008521.mm.1 | NM_026309.2 | Refseq |
Mouse Expression Set 430 | 1448536_at | - | - |
Mouse Expression Set 430 | 1448536_PM_at | - | - |
Murine Genome U74v2 Set | 94455_PM_at | - | - |
Murine Genome U74v2 Set | 94455_at | - | - |
Mouse Genome 430A 2.0 Array | 1448536_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448536_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448536_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448536_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 580691 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 780164 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000780164 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252710 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1252710 | - | - |
mouse-6 v1.1 (Illumina) | scl067678.4_130-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 780164 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl067678.4_130-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 780164 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2837865 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2837865 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067678.4_130-S | - | - |
