Assay Details
| Gene Symbol: Pbdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281871.1 | NP_001268800.1 | 3 - 4 | 199 | 97 | |
| NM_026312.5 | NP_080588.1 | 3 - 4 | 199 | 97 | ||
| GenBank mRNA | AK007655.1 | - | 3 - 4 | 181 | 97 | |
| AK007872.1 | - | 2 - 3 | 68 | 97 | ||
| AK011610.1 | - | 3 - 4 | 163 | 97 | ||
| AK030507.1 | - | 3 - 4 | 181 | 97 | ||
| AK076889.1 | - | 3 - 4 | 181 | 97 | ||
| BC035042.1 | - | 3 - 4 | 163 | 1224959 | 97 | |
| BC044719.1 | - | 3 - 4 | 147 | 6394829 | 97 | |
| BC061070.1 | - | 3 - 4 | 160 | 6594922 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pbdc1
Genomic Map