Assay Details
| Gene Symbol: Prss37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026317.2 | NP_080593.1 | 1 - 2 | 104 | 86 | |
| GenBank mRNA | AK006028.1 | - | 1 - 2 | 108 | 86 | |
| AK016088.1 | - | 1 - 2 | 109 | 86 | ||
| AK029780.1 | - | 1 - 2 | 81 | 86 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK006028.1,AK016088.1,AK029780.1,NM_026317.2 transcripts.
Set Membership: |
|

Prss37
Genomic Map