Assay Details
| Gene Symbol: Prss37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026317.2 | NP_080593.1 | 3 - 4 | 401 | 68 | |
| GenBank mRNA | AK006028.1 | - | 3 - 4 | 405 | 68 | |
| AK016088.1 | - | 3 - 4 | 406 | 68 | ||
| AK029780.1 | - | 3 - 4 | 378 | 68 | ||
| BC125467.1 | - | 2 - 3 | 264 | 40129982 | 68 | |
| BC125493.1 | - | 2 - 3 | 264 | 40130008 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prss37
Genomic Map