Assay Details
| Gene Symbol: Ilf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026374.3 | NP_080650.1 | 10 - 11 | 785 | 75 | |
| GenBank mRNA | AK012364.1 | - | 10 - 11 | 772 | 75 | |
| AK013858.1 | - | 10 - 11 | 831 | 75 | ||
| AK078003.1 | - | 10 - 11 | 772 | 75 | ||
| AK135449.1 | - | 10 - 11 | 860 | 75 | ||
| AK135539.1 | - | 10 - 11 | 772 | 75 | ||
| AK157129.1 | - | 10 - 11 | 840 | 75 | ||
| BC004033.1 | - | 2 - 3 | 152 | 75 | ||
| BC024718.1 | - | 10 - 11 | 785 | 5012365 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ilf2
Genomic Map