Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800020012.mm.1 |
NM_026399.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020012.mm.1 |
NM_026399.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020012.mm.1 |
TC0800002603.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020011.mm.1 |
NM_026399.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020011.mm.1 |
NM_026399.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020011.mm.1 |
TC0800002603.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020016.mm.1 |
NM_026399.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020016.mm.1 |
NM_026399.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020016.mm.1 |
TC0800002603.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020013.mm.1 |
NM_026399.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020013.mm.1 |
NM_026399.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020013.mm.1 |
TC0800002603.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020017.mm.1 |
NM_026399.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020017.mm.1 |
NM_026399.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020017.mm.1 |
TC0800002603.mm.1 |
MTA TC |