Assay Details
Gene Symbol: Galntl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026449.3 | NP_080725.2 | 4 - 5 | 678 | 152 | |
GenBank mRNA | AK005605.1 | - | 4 - 5 | 699 | 152 | |
AK006193.1 | - | 4 - 5 | 657 | 152 | ||
AK016415.1 | - | 4 - 5 | 679 | 152 | ||
BC049554.1 | - | 4 - 5 | 660 | 152 | ||
BC100447.1 | - | 4 - 5 | 531 | 152 | ||
BC145757.1 | - | 4 - 5 | 538 | 40131060 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
