Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100035703.mm.1 |
NM_026542.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035703.mm.1 |
NM_026542.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035703.mm.1 |
TC1100003836.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100035707.mm.1 |
NM_026542.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035707.mm.1 |
NM_026542.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035707.mm.1 |
TC1100003836.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100035709.mm.1 |
NM_026542.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035709.mm.1 |
NM_026542.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035709.mm.1 |
TC1100003836.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100035706.mm.1 |
NM_026542.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035706.mm.1 |
NM_026542.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035706.mm.1 |
TC1100003836.mm.1 |
MTA TC |