Assay Details
| Gene Symbol: Arpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026552.3 | NP_080828.1 | 1 - 2 | 95 | 65 | |
| GenBank mRNA | AK017993.1 | - | 1 - 2 | 96 | 65 | |
| AK018069.1 | - | 1 - 2 | 33 | 65 | ||
| AK039215.1 | - | 1 - 2 | 43 | 65 | ||
| AK133626.1 | - | 1 - 2 | 43 | 65 | ||
| AK136082.1 | - | 1 - 2 | 38 | 65 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK017993.1,AK018069.1,AK039215.1,AK133626.1,AK136082.1,NM_026552.3 transcripts.
Set Membership: |
|

Arpc4
Genomic Map