Assay Details
| Gene Symbol: Arpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026552.3 | NP_080828.1 | 2 - 3 | 212 | 81 | |
| GenBank mRNA | AJ278129.1 | - | 2 - 3 | 138 | 81 | |
| AK017993.1 | - | 2 - 3 | 213 | 81 | ||
| AK018069.1 | - | 2 - 3 | 150 | 81 | ||
| AK039215.1 | - | 2 - 3 | 160 | 81 | ||
| AK076248.1 | - | 2 - 3 | 160 | 81 | ||
| AK077325.1 | - | 2 - 3 | 134 | 81 | ||
| AK133626.1 | - | 2 - 3 | 160 | 81 | ||
| AK136082.1 | - | 2 - 3 | 155 | 81 | ||
| AK150690.1 | - | 2 - 3 | 146 | 81 | ||
| AK153380.1 | - | 2 - 3 | 144 | 81 | ||
| AK166440.1 | - | 2 - 3 | 134 | 81 | ||
| AK193553.1 | - | 1 - 2 | 83 | 81 | ||
| AK194784.1 | - | 1 - 2 | 83 | 81 | ||
| AK197517.1 | - | 1 - 2 | 83 | 81 | ||
| BC015280.1 | - | 1 - 2 | 124 | 81 | ||
| BC055309.1 | - | 1 - 2 | 123 | 6529255 | 81 | |
More Information
Set Membership: |
|

Arpc4
Genomic Map