Assay Details
| Gene Symbol: 4930591A17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026596.2 | NP_080872.1 | 2 - 3 | 323 | 83 | |
| GenBank mRNA | AK019836.1 | - | 2 - 3 | 316 | 83 | |
| AK019837.1 | - | 2 - 3 | 326 | 83 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245102 | AK019837.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245102 | AK019836.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245102 | AK019837.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245102 | AK019836.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245102 | AK019837.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245102 | AK019836.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245102 | AK019837.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245102 | AK019836.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245102 | AK019837.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245102 | AK019836.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245102 | AK019837.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245102 | AK019836.1 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01751246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01227365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01174713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01174720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01240082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01174707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01238088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01240083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01361195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01240079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01240085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01998790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01262258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01240080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02237302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01723906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01361190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01739978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01240071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01240078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01163041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01751243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01240074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01361196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21896817 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1450305_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450305_at | - | - |
| Mouse Genome 430 2.0 Array | 1450305_PM_at | - | - |
| Mouse Expression Set 430 | 1450305_at | - | - |
| Mouse Expression Set 430 | 1450305_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450305_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450305_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450305_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450305_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 695034 | - | - |

4930591A17Rik
Genomic Map