Assay Details
| Gene Symbol: Gtsf1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026630.2 | NP_080906.1 | 1 - 2 | 59 | 94 | |
| GenBank mRNA | AK019131.1 | - | 1 - 2 | 60 | 94 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK019131.1,NM_026630.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810983 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810977 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810973 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810974 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810969 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810981 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810980 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810982 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810970 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P488501 | BC048439.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P488501 | NM_026630.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P488501 | AK018886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P488501 | AK019131.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P488501 | AK160150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P488501 | BC048439.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P488501 | NM_026630.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P488501 | AK018886.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P488501 | AK019131.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P488501 | AK160150.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183608 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136677 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P488501 | BC048439.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P488501 | NM_026630.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P488501 | AK018886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P488501 | AK019131.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P488501 | AK160150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P488501 | BC048439.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P488501 | NM_026630.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P488501 | AK018886.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P488501 | AK019131.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P488501 | AK160150.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136677 | - | - |
| Whole Mouse Genome G4122A | A_52_P488501 | BC048439.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488501 | NM_026630.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488501 | AK018886.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488501 | AK019131.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488501 | AK160150.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488501 | BC048439.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488501 | NM_026630.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488501 | AK018886.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488501 | AK019131.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488501 | AK160150.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128890 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136677 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810977 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810969 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810982 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810983 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810981 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810974 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810970 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810973 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21810980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21810378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810404 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01772745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01891873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02003987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01751501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01403465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02125700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01781162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02235244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01891872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21810384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02235243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01403468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01870044 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10489352 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489351 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489350 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418994_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10489349 | - | - |
| Mouse Genome 430 2.0 Array | 1418994_at | - | - |
| Mouse Genome 430 2.0 Array | 1418994_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68236_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68236_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200043390.mm.1 | NM_026630.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043390.mm.1 | NM_026630.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043390.mm.1 | TC0200005087.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200043391.mm.1 | NM_026630.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043391.mm.1 | NM_026630.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043391.mm.1 | TC0200005087.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200043393.mm.1 | NM_026630.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043393.mm.1 | NM_026630.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043393.mm.1 | TC0200005087.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418994_PM_at | - | - |
| Mouse Expression Set 430 | 1418994_at | - | - |
| Murine Genome U74v2 Set | 169104_r_at | - | - |
| Murine Genome U74v2 Set | 169104_PM_r_at | - | - |
| Murine Genome U74v2 Set | 166732_PM_f_at | - | - |
| Murine Genome U74v2 Set | 166732_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418994_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418994_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 853937 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1660129 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001660129 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2732954 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2732954 | - | - |
| mouse-6 v1.1 (Illumina) | scl18388.2.4_45-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1660129 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18388.2.4_45-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660129 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2732954 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2980015 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2980015 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2732954 | - | - |

Gtsf1l
Genomic Map