Assay Details
| Gene Symbol: Fam111a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026640.2 | NP_080916.1 | 3 - 4 | 343 | 147 | |
| GenBank mRNA | AK018830.1 | - | 3 - 4 | 352 | 147 | |
| AK019499.1 | - | 3 - 4 | 344 | 147 | ||
| AK045189.1 | - | 3 - 4 | 345 | 147 | ||
| AK172402.1 | - | 3 - 4 | 356 | 147 | ||
| BC038020.1 | - | 3 - 4 | 337 | 3599082 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394802 | BC019638.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394802 | BC038020.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394802 | AK142496.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394802 | AK172402.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394802 | AK019499.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394802 | BC019638.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394802 | BC038020.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394802 | AK142496.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394802 | AK172402.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394802 | AK019499.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 167835 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318266 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655961 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451186 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701689 | AF025653.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701689 | AF034568.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701689 | AK002922.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701689 | AK079488.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701689 | AK142016.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701689 | AK150497.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701689 | AK151463.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701689 | AK160007.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701689 | AK160706.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701689 | AK169305.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701689 | BC031526.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701689 | BC043657.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701689 | NM_011884.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313671 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168231 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19851 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158776 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159497 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142082 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394802 | BC019638.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394802 | BC038020.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394802 | AK142496.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394802 | AK172402.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394802 | AK019499.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394802 | BC019638.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394802 | BC038020.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394802 | AK142496.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394802 | AK172402.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394802 | AK019499.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31961405 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142082 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159497 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09108807 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158776 | - | - |
| Whole Mouse Genome G4122A | A_51_P394802 | BC019638.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394802 | BC038020.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394802 | AK142496.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394802 | AK172402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394802 | AK019499.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394802 | BC019638.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394802 | BC038020.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394802 | AK142496.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394802 | AK172402.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394802 | AK019499.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120588 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158776 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159497 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142082 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14249136 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14249135 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14249138 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14238174 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14249133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31961462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14211444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14211447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14145353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14249139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14234227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14167821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14414617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14414616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14202948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14202945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14322955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14249142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31961410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14468256 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10461726 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461727 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422628_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461723 | - | - |
| Mouse Genome 430 2.0 Array | 1422628_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422628_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107373_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107373_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900002920.mm.1 | NM_026640.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900002920.mm.1 | NM_026640.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900002920.mm.1 | TC1900000247.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422628_PM_at | - | - |
| Mouse Expression Set 430 | 1422628_at | - | - |
| Murine Genome U74v2 Set | 161142_PM_at | - | - |
| Murine Genome U74v2 Set | 161142_at | - | - |
| Murine Genome U74v2 Set | 171579_r_at | - | - |
| Murine Genome U74v2 Set | 171579_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422628_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422628_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422628_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422628_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 627512 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4760369 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4760369 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004760369 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2718843 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2718843 | - | - |
| mouse-6 v1.1 (Illumina) | scl53341.4_558-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4760369 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53341.4_558-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4760369 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2718843 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2718843 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl53341.4_558-S | - | - |

Fam111a
Genomic Map