Assay Details
| Gene Symbol: Mfsd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026660.2 | NP_080936.1 | 11 - 12 | 1371 | 76 | |
| GenBank mRNA | AK003763.1 | - | 6 - 7 | 639 | 76 | |
| AK018736.1 | - | 11 - 12 | 1384 | 76 | ||
| AK089055.1 | - | 11 - 12 | 1388 | 76 | ||
| AK144407.1 | - | 2 - 3 | 91 | 76 | ||
| AK152438.1 | - | 11 - 12 | 1357 | 76 | ||
| BC023012.1 | - | 11 - 12 | 1371 | 5376230 | 76 | |
| BC064775.1 | - | 11 - 12 | 1269 | 30023877 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd10
Genomic Map