Assay Details
| Gene Symbol: Prps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026662.4 | NP_080938.1 | 4 - 5 | 645 | 104 | |
| GenBank mRNA | AK012819.1 | - | 4 - 5 | 677 | 104 | |
| AK019169.2 | - | 4 - 5 | 614 | 104 | ||
| AK029690.1 | - | 4 - 5 | 660 | 104 | ||
| AK045007.1 | - | 4 - 5 | 646 | 104 | ||
| BC024942.1 | - | 4 - 5 | 632 | 4022452 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prps2
Genomic Map