Assay Details
Gene Symbol: Calml3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027416.3 | NP_081692.1 | 1 | 1225 | 77 | |
GenBank mRNA | AK010118.1 | - | 1 | 1222 | 77 | |
BX516975.1 | - | 1 | 401 | 77 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137709 | AK010118.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137709 | NM_027416.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137709 | BB144976.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137709 | BC005457.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137709 | AK010118.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137709 | NM_027416.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137709 | BB144976.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P137709 | BC005457.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083317 | AK010118.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083317 | NM_027416.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083317 | BC005457.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932871 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155840 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083317 | AK010118.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083317 | NM_027416.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083317 | BC005457.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137709 | AK010118.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137709 | NM_027416.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137709 | BB144976.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137709 | BC005457.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137709 | AK010118.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137709 | NM_027416.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137709 | BB144976.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P137709 | BC005457.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789917 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789907 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789920 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789903 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789891 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789902 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789889 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789906 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789923 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789925 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789890 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789884 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789883 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789928 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789908 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789913 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789899 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789927 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789894 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789914 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789893 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789929 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789911 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789892 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789921 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789919 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789915 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789896 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789886 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28789904 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155840 | - | - |
Whole Mouse Genome G4122A | A_51_P137709 | AK010118.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137709 | NM_027416.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137709 | BB144976.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137709 | BC005457.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137709 | AK010118.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137709 | NM_027416.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137709 | BB144976.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P137709 | BC005457.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107103 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155840 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10864877 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10864871 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10864857 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10864842 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10864862 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10864861 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10864858 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10864874 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10864865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789881 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28789915 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10980887 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10982966 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789854 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789932 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789896 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789871 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789819 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789899 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10694622 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789889 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789880 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789893 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789906 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789913 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789861 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10982969 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10460219 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789867 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789915 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789903 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789852 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789882 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789892 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789927 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789849 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789874 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789858 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10694621 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789865 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789917 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789911 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789847 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789928 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789821 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10864858 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789931 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789933 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789877 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789881 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789851 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789925 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10565597 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789879 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789919 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789891 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789857 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789890 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789886 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789875 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789920 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789914 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789929 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789904 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789818 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789820 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789863 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10982971 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789885 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789870 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11039958 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28789894 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10407419 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10407417 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10407418 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418608_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10407416 | - | - |
Mouse Genome 430 2.0 Array | 1418608_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418608_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70405_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70405_PM_at | - | - |
Mouse Expression Set 430 | 1418608_at | - | - |
Mouse Expression Set 430 | 1418608_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418608_at | - | - |
Mouse Genome 430A 2.0 Array | 1418608_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418608_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418608_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 442843 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3360021 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003360021 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2592901 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2592901 | - | - |
mouse-6 v1.1 (Illumina) | scl44317.1.1_34-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3360021 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl44317.1.1_34-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360021 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2592901 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2592901 | - | - |
