Assay Details
| Gene Symbol: Eri2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027698.5 | NP_081974.3 | 5 - 6 | 583 | 98 | |
| GenBank mRNA | AK015831.1 | - | 493 | 98 | ||
| AK016902.1 | - | 493 | 98 | |||
| AK029056.1 | - | 5 - 6 | 508 | 98 | ||
| AK129376.1 | - | 4 - 5 | 400 | 98 | ||
| AK135821.1 | - | 5 - 6 | 496 | 98 | ||
| AK137621.1 | - | 5 - 6 | 529 | 98 | ||
| AK141507.1 | - | 5 - 6 | 504 | 98 | ||
| AK160698.1 | - | 5 - 6 | 479 | 98 | ||
| AK165163.1 | - | 5 - 6 | 493 | 98 | ||
| BC052150.1 | - | 1210 | 98 | |||
| BC090961.1 | - | 5 - 6 | 485 | 30648850 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eri2
Genomic Map