Assay Details
| Gene Symbol: Chn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113246.2 | NP_001106717.2 | 9 - 10 | 1195 | 71 | |
| NM_029716.3 | NP_083992.1 | 2 - 3 | 396 | 71 | ||
| NM_175752.3 | NP_786928.2 | 3 - 4 | 1007 | 71 | ||
| GenBank mRNA | AB264774.1 | - | 1 - 2 | 797 | 71 | |
| AF332069.1 | - | 3 - 4 | 515 | 71 | ||
| AF332070.1 | - | 3 - 4 | 515 | 71 | ||
| AK007182.1 | - | 2 - 3 | 396 | 71 | ||
| AK032630.1 | - | 3 - 4 | 965 | 71 | ||
| AK049943.1 | - | 3 - 4 | 951 | 71 | ||
| AK075606.1 | - | 3 - 4 | 969 | 71 | ||
| AK147686.1 | - | 3 - 4 | 951 | 71 | ||
| BC010825.1 | - | 3 - 4 | 982 | 4219273 | 71 | |
| BC024796.1 | - | 3 - 4 | 958 | 5360438 | 71 | |
| BC025023.1 | - | 3 - 4 | 938 | 5364554 | 71 | |
| BC054770.1 | - | 9 - 10 | 947 | 6415204 | 71 | |
More Information
Set Membership: |
|

Chn1
Genomic Map