Assay Details
| Gene Symbol: Rab27b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB067775.1 | - | 4 - 5 | 601 | 77 | |
| AB120512.1 | - | 3 - 4 | 345 | 77 | ||
| AB232623.1 | - | 3 - 4 | 345 | 77 | ||
| AF328893.1 | - | 4 - 5 | 614 | 77 | ||
| AK049854.1 | - | 5 - 6 | 498 | 77 | ||
| AY010898.1 | - | 3 - 4 | 345 | 77 | ||
| BC018443.1 | - | 4 - 5 | 581 | 3601865 | 77 | |
| BC032928.1 | - | 4 - 5 | 534 | 5120275 | 77 | |
| BC096496.1 | - | 5 - 6 | 1010 | 30738460 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab27b
Genomic Map