Assay Details
| Gene Symbol: Siglece | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031181.2 | NP_112458.2 | 4 - 5 | 1057 | 69 | |
| GenBank mRNA | AY348552.1 | - | 4 - 5 | 1066 | 69 | |
| AY371487.1 | - | 4 - 5 | 1046 | 69 | ||
| BC023280.1 | - | 4 - 5 | 1056 | 3662856 | 69 | |
More Information
Set Membership: |
|

Siglece
Genomic Map