Assay Details
| Gene Symbol: Bicc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031397.2 | NP_113574.1 | 6 - 7 | 626 | 70 | |
| GenBank mRNA | AF319464.1 | - | 6 - 7 | 627 | 70 | |
| AK076154.1 | - | 6 - 7 | 679 | 70 | ||
| AK089066.1 | - | 6 - 7 | 457 | 70 | ||
| BC111523.2 | - | 6 - 7 | 627 | 40048106 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bicc1
Genomic Map