Assay Details
| Gene Symbol: Smarcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031842.2 | NP_114030.2 | 2 - 3 | 399 | 95 | |
| GenBank mRNA | BC026783.1 | - | 2 - 3 | 311 | 4481097 | 95 |
| BC059921.1 | - | 2 - 3 | 369 | 95 | ||
| BC079839.1 | - | 2 - 3 | 345 | 6810462 | 95 | |
| CV562378.1 | - | 2 - 3 | 399 | 95 | ||
| M25773.1 | - | 2 - 3 | 355 | 95 | ||
| U66620.1 | - | 2 - 3 | 355 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smarcd1
Genomic Map