Assay Details
| Gene Symbol: Enpp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168620.1 | NP_001162091.1 | 2 - 3 | 908 | 74 | |
| NM_032003.2 | NP_114392.1 | 2 - 3 | 994 | 74 | ||
| GenBank mRNA | AF233377.1 | - | 2 - 3 | 971 | 74 | |
| AK032341.1 | - | 2 - 3 | 897 | 74 | ||
| AK220510.1 | - | 2 - 3 | 937 | 74 | ||
| BC089320.1 | - | 3 - 4 | 714 | 74 | ||
| BC138977.1 | - | 2 - 3 | 909 | 8861999 | 74 | |
| BC138978.1 | - | 2 - 3 | 909 | 8862000 | 74 | |
| BC145484.1 | - | 2 - 3 | 909 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Enpp5
Genomic Map