Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700025517.mm.1 |
TC0700002854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025517.mm.1 |
NM_033080.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025517.mm.1 |
NM_033080.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025521.mm.1 |
TC0700002854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025521.mm.1 |
NM_033080.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025521.mm.1 |
NM_033080.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025519.mm.1 |
TC0700002854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025519.mm.1 |
NM_033080.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025519.mm.1 |
NM_033080.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025520.mm.1 |
TC0700002854.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025520.mm.1 |
NM_033080.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025520.mm.1 |
NM_033080.2 |
Refseq |