Assay Details
Gene Symbol: Actn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033268.4 | NP_150371.4 | 15 - 16 | 2075 | 69 | |
GenBank mRNA | AF248643.1 | - | 15 - 16 | 1843 | 69 | |
AY036877.1 | - | 16 - 17 | 2019 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
