Assay Details
| Gene Symbol: P2rx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033321.3 | NP_201578.2 | 3 - 4 | 429 | 66 | |
| GenBank mRNA | AF333331.1 | - | 3 - 4 | 435 | 66 | |
| AK134662.1 | - | 3 - 4 | 389 | 66 | ||
| BC138189.1 | - | 3 - 4 | 429 | 8861208 | 66 | |
| BC145181.1 | - | 3 - 4 | 429 | 9053732 | 66 | |
| JX570808.1 | - | 3 - 4 | 425 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P2rx5
Genomic Map