Assay Details
| Gene Symbol: P2rx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033321.3 | NP_201578.2 | 8 - 9 | 955 | 64 | |
| GenBank mRNA | AF333331.1 | - | 8 - 9 | 961 | 64 | |
| AK134662.1 | - | 8 - 9 | 915 | 64 | ||
| BC138189.1 | - | 8 - 9 | 955 | 8861208 | 64 | |
| BC145181.1 | - | 8 - 9 | 955 | 9053732 | 64 | |
| JX570808.1 | - | 7 - 8 | 1218 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P2rx5
Genomic Map